Saint-Gobain™ Tygon S3™ Clear Tubing, Formula B-44-4X, Non-DEHP
Specially designed to ensure bacterial-free fluid path for food processing applications
Manufacturer: Saint-Gobain™ AAA00029
 View more versions of this product
Catalog No. 14-171-156
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Color Clear
    Max. Pressure 1379psi
    Diameter (English) Outer 5/8 in.
    Diameter (English) Inner 3/8 in.
  • Description & Specifications
Related Products
Product Content Feedback