Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain™ Tygon S3™ Clear Tubing, Formula B-44-4X

Manufacturer:  Saint-Gobain™ AAA00002

 View more versions of this product

Catalog No. 14-171-140


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Tygon
0.12 in.
0.03 in.
3.2mm
Clear
50 ft.
60ψ
74°C
0.8mm
6.35mm
Shore A: 65
Yes
0.06 in.
1.6mm
FDA 21 CFR, 175.300, 3-A and NSF Standard 51
Aseptic filling, condiment dispensing, dairy processing, vitamin and flavor concentrate systems, soft-serve dispensing
15.2m
165°F
-45 to 79°C
390%
29.9 in. Hg (at 73°F)
0.25 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit