Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion
Manufacturer: Saint-Gobain™ ACF00009
 View more versions of this product
Catalog No. 14-171-213
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Material Non-DEHP; Phthalate-free; Bio-based; Chemical resistant
    Color Clear
    Max. Pressure Max. working: 17psi at 73 deg.F
    Thickness 1/32 in.
    Diameter (English) Outer 7/32 in.
    Diameter (English) Inner 5/32 in.
  • Description & Specifications
Related Products
Product Content Feedback