Learn More
Saint-Gobain Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion
$56.00 - $1259.00
Specifications
Color Clear
Material Tygon S3 E-3603
View More Specs
Products 15
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Thickness (English) Price Quantity & Availability  
14-171-209
View Documents
Saint-Gobain
ACF00003
1/16 in. 3/16 in. 69 psi at 73 deg. F 1/16 in. Pack of 50 for $67.70
Add to cart
14-171-210
View Documents
Saint-Gobain
ACF00005
1/16 in. 3/32 in. 47 psi at 73 deg.F 7/32 in. Pack of 50 for $91.10
Add to cart
14-171-213
View Documents
Saint-Gobain
ACF00009
5/32 in. 7/32 in. 17 psi at 73 deg.F 1/32 in. Pack of 50 for $56.00
Add to cart
14-171-214
View Documents
Saint-Gobain
ACF00010
5/32 in. 9/32 in. 30 psi at 73 deg.F 1/16 in. Pack of 50 for $119.30
Add to cart
14-171-217
View Documents
Saint-Gobain
ACF00013
3/16 in. 3/8 in. 36 psi at 73 deg.F 3/32 in. Pack of 50 for $252.00
Add to cart
14-171-219
View Documents
Saint-Gobain
ACF00016
1/4 in. 5/16 in. 12 psi at 73 deg.F 1/32 in. Pack of 50 for $78.40
Add to cart
14-171-226
View Documents
Saint-Gobain
ACF00025
5/16 in. 5/8 in. 36 psi at 73 deg.F 5/32 in. Pack of 50 for $449.00
Add to cart
14-171-229
View Documents
Saint-Gobain
ACF00029
3/8 in. 5/8 in. 13 psi at 73 deg.F 1/8 in. Pack of 50 for $489.06
Add to cart
14-171-231
View Documents
Saint-Gobain
ACF00032
7/16 in. 9/16 in. 13 psi at 73 deg.F 1/16 in. Pack of 50 for $281.00
Add to cart
14-171-232
View Documents
Saint-Gobain
ACF00033
7/16 in. 5/8 in. 18 psi at 73 deg.F 3/32 in. Pack of 50 for $341.50
Add to cart
14-171-241
View Documents
Saint-Gobain
ACF00045
5/8 in. 13/16 in. 13 psi at 73 deg. F 3/32 in. Pack of 50 for $500.00
Add to cart
14-171-248
View Documents
Saint-Gobain
ACF00057
3/4 in. 1-1/4 in. 25 psi at 73deg.F 1/4 in. Pack of 50 for $1,259.00
Add to cart
14-171-250
View Documents
Saint-Gobain
ACF00059
7/8 in. 1-1/8 in. 36 psi at 73 deg.F 1/8 in. Pack of 50 for $639.50
Add to cart
14-171-255
View Documents
Saint-Gobain
ACF00069
1-1/4 in. 1-1/2 in. 13 psi at 73 deg.F 1/8 in. Pack of 50 for $895.00
Add to cart
14-171-237
View Documents
Saint-Gobain
ACF00039
1/2 in. 13/16 in. 24 psi at 73 deg.F 5/32 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Bio-based formulation and contain no BPA or phthalates
  • Provide high chemical resistance
  • Non-oxidizing and non-contaminating
  • Smooth, polished inner wall helps prevent buildup to facilitate cleaning
  • Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Extra-heavy walls withstand full vacuum at room temperature (29.9 in. [759mm] of mercury at 23 °C [73 °F] and up to 27 in. [686mm] of mercury at 60°C [140°F])
  • Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
  • Instrumentation connection, vent, drain and other general laboratory applications
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Condensers
  • Incubators
  • Desiccators
  • Gas lines
  • Drain lines
Compliance
FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.
Specifications
Specifications
Clear
Tygon S3 E-3603