Learn More
Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion
Manufacturer:  Saint-Gobain™ ACF00025
 View more versions of this product
Catalog No. 14-171-226
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Non-DEHP; Phthalate-free; Bio-based; Chemical resistant
5/16 in.
Max. working: 36psi at 73 deg.F
5/8 in.
Clear
5/32 in.