Learn More
Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes handles virtually all inorganic chemicals found in the lab.
Manufacturer:  Saint-Gobain™ ACF00020
 View more versions of this product
Catalog No. 14-171-222
  Check Availability
Add to cart
Specifications
Description
  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food & beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
Description & Specifications
Specifications
Non-DEHP; Phthalate-free; Bio-based; Chemical resistant
1/4 in.
Max. working: 53psi at 73 deg.F
5/8 in.
Clear
3/16 in.