Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes
Formulated for resistance to flex-fatigue and abrasion
Manufacturer: Saint-Gobain™ ACF00040
 View more versions of this product
Catalog No. 14-171-238
  Check Availability
Add to Cart
    Specifications
    Material Non-DEHP; Phthalate-free; Bio-based; Chemical resistant;
    Color Clear
    Max. Pressure Max. working: 36psi at 73 deg.F
    Thickness 5/16 in.
    Diameter (English) Outer 1-1/8 in.
    Diameter (English) Inner 1/2 in.
  • Description & Specifications
Related Products
Product Content Feedback