Learn More
Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes handles virtually all inorganic chemicals found in the lab.
Manufacturer:  Saint-Gobain™ ACF00049
 View more versions of this product
Catalog No. 14-171-244
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Non-DEHP; Phthalate-free; Bio-based; Chemical resistant
5/8 in.
Max. working: 43psi at 73 deg.F
1-3/8 in.
Clear
3/8 in.