Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon S3™ E-3603 Flexible Tubings, Vacuum Sizes handles virtually all inorganic chemicals found in the lab.
Manufacturer: Saint-Gobain™ ACF00015
 View more versions of this product
Catalog No. 14-171-218
  Check Availability
Add to Cart
Related Products
Product Content Feedback