Learn More
Saint-Gobain™ Tygon™ Ultra-Chemical-Resistant Tubing, Formula 2375
Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.
Manufacturer:  Saint-Gobain™ AJK00012
 View more versions of this product
Catalog No. 14-169-22C
  Check Availability
Add to cart
Specifications
Description
  • Chemical-resistant tubing for food and beverage applications
  • Virtually unaffected by acids, bases, ketones, salts and alcohols
  • Outstanding chemical resistance
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Safer disposal - Releases no harmful and corrosive hydrochloride gas
  • Smoother inner surface - Provides better flow and inhibits particulate buildup
  • Low sorption - Minimizes cross-contamination
  • Clear tubing for easier and better observation
Description & Specifications
Specifications
Tygon
0.188 in.
4.8mm
50 ft.
1.6mm
0.3125 in.
0.0625 in.
7.9mm
15m