Learn More
Saint-Gobain Tygon™ Ultra-Chemical-Resistant Tubing, Formula 2375
Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.
$140.10 - $357.50
Specifications
Material Tygon
View More Specs
Products 9
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (Metric) Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (Metric) Thickness (English) Price Quantity & Availability  
14-169-22A
View Documents
Saint-Gobain
AJK00003
0.063 in. 3/16 in. 1.6 mm 0.0625 in.
Pack for $140.10
Add to cart
14-169-22D
View Documents
Saint-Gobain
AJK00017
1/4 in. 3/8 in. 1.6 mm 0.0625 in.
Pack for $314.50
Add to cart
14-169-22C
View Documents
Saint-Gobain
AJK00012
0.188 in. 5/16 in. 1.6 mm 0.0625 in.
Pack for $251.00
Add to cart
14-169-22B
View Documents
Saint-Gobain
AJK00007
0.13 in. 1/4 in. 1.6 mm 0.0625 in.
Pack for $236.00
Add to cart
14-169-22E
View Documents
Saint-Gobain
AJK00022
5/16 in. 7/16 in. 1.6 mm 0.0625 in.
Pack for $307.00
Add to cart
14-169-22F
View Documents
Saint-Gobain
AJK00027
3/8 in. 1/2 in. 1.6 mm 0.0625 in.
Pack for $357.50
Add to cart
14-169-22G
View Documents
Saint-Gobain
AJK00038
1/2 in. 3/4 in. 3.2 mm 0.125 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-600-1B
View Documents
Saint-Gobain
AJK42064
1 in. 1.38 in. 4.8 mm 0.19 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-169-22H
View Documents
Saint-Gobain
AJK00046
5/8 in. 7/8 in. 3.2 mm 0.125 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.
  • Chemical-resistant tubing for food and beverage applications
  • Virtually unaffected by acids, bases, ketones, salts and alcohols
  • Outstanding chemical resistance
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Safer disposal - Releases no harmful and corrosive hydrochloride gas
  • Smoother inner surface - Provides better flow and inhibits particulate buildup
  • Low sorption - Minimizes cross-contamination
  • Clear tubing for easier and better observation
Specifications
Specifications
Tygon