Learn More
Saint-Gobain Tygothane™ Polyurethane Pressure Tubing
$1145.00 - $1573.00
Specifications
Material Polyurethane C-544-A I.B.
For Use With (Application) Resistant to chemicals, oils, greases and solvents
For Use With Laboratory pumps and equipment
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (English) Inner Overall Length (English) Maximum Pressure Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Overall Length (English) Maximum Pressure Thickness (English) Price Quantity & Availability  
02-589-1H
missing translation for 'viewDocuments'
Saint-Gobain
AZY00074
1-1/2 in. 50 ft. 80 psi 0.25 in.
Pack of 50 for $1,573.00
Add to cart
02-589-1E
missing translation for 'viewDocuments'
Saint-Gobain
AZY02054
3/4 in. 100 ft. 150 psi 0.16 in.
Pack of 100 for $1,145.00
Add to cart
02-589-1F
missing translation for 'viewDocuments'
Saint-Gobain
AZY02064
1 in. 100 ft. 120 psi 0.19 in.
Pack of 100 for $1,451.00
Add to cart
02-589-1C
missing translation for 'viewDocuments'
Saint-Gobain
AZY02038
1/2 in. 100 ft. 195 psi 0.125 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-589-1A
missing translation for 'viewDocuments'
Saint-Gobain
AZY02019
1/4 in. 100 ft. 275 psi 0.125 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Specifications
Specifications
Polyurethane C-544-A I.B.
Resistant to chemicals, oils, greases and solvents
Laboratory pumps and equipment