Learn More
Saint-Gobain™ Tygothane™ Polyurethane Pressure Tubing
Manufacturer:  Saint-Gobain™ AZY02019
 View more versions of this product
Catalog No. 02-589-1A
  Check Availability
Add to cart
Specifications
Specifications
1/4 in.
1/2 in.
0.125 in.
Resistant to chemicals, oils, greases and solvents
Polyurethane C-544-A I.B.
100 ft.
Laboratory pumps and equipment
275psi