Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Saint-Gobain™ Tygothane™ Polyurethane Pressure Tubing
Manufacturer:  Saint-Gobain™ AZY02038
 View more versions of this product
Catalog No. 02-589-1C
Specifications
Specifications
1/2 in.
3/4 in.
0.125 in.
Resistant to chemicals, oils, greases and solvents
Polyurethane C-544-A I.B.
100 ft.
Laboratory pumps and equipment
195psi