Learn More
Saint-Gobain™ Tygothane™ Polyurethane Pressure Tubing
Manufacturer:  Saint-Gobain™ AZY02064
 View more versions of this product
Catalog No. 02-589-1F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyurethane C-544-A I.B.
1 in.
1 in.
Resistant to chemicals, oils, greases and solvents
100 ft.
0.19 in.
1.375 in.
0.19 in.
1.375 in.
Laboratory pumps and equipment
120psi