Learn More
Saint-Gobain™ Tygothane™ Polyurethane Pressure Tubing
Manufacturer:  Saint-Gobain™ AZY00074
 View more versions of this product
Catalog No. 02-589-1H
Add to cart
Specifications
Specifications
1-1/2 in.
2 in.
0.25 in.
Resistant to chemicals, oils, greases and solvents
Polyurethane C-544-A I.B.
50 ft.
Laboratory pumps and equipment
80psi