Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldehyde, 98%, Spectrum™

Click to view available options
Quantity:
250 mL
Packaging:
Amber Glass Bottle

SA122, 90-02-8, C7H6O2

Spectrum™ Salicylaldehyde , also known as Salicylic aldehyde or 2-Hydroxybenzaldehyde, is a precursor to a several types of commercially important chelating agents. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Packaging Amber Glass Bottle
Refractive Index 1.571-1.575
Quantity 250 mL
CAS 90-02-8
CAS Max % 1
Molecular Formula C7H6O2
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1C=O
IUPAC Name 2-hydroxybenzaldehyde
Molecular Weight (g/mol) 122.12
Percent Purity 98%
Grade Reagent
Show More Show Less