Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldehyde, 99%, ACROS Organics™

Click to view available options
Quantity:
2g
100g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
90-02-8,108-95-2
C7H6O2
122.12
SMQUZDBALVYZAC-UHFFFAOYSA-N
salicylaldehyde,o-hydroxybenzaldehyde,o-formylphenol,salicylal,2-formylphenol,benzaldehyde, 2-hydroxy,salicylic aldehyde,salicyladehyde,2-hydroxy-benzaldehyde,benzaldehyde, o-hydroxy
6998
CHEBI:16008
2-hydroxybenzaldehyde
C1=CC=C(C(=C1)C=O)O
90-02-8,108-95-2
C7H6O2
122.12
SMQUZDBALVYZAC-UHFFFAOYSA-N
salicylaldehyde,o-hydroxybenzaldehyde,o-formylphenol,salicylal,2-formylphenol,benzaldehyde, 2-hydroxy,salicylic aldehyde,salicyladehyde,2-hydroxy-benzaldehyde,benzaldehyde, o-hydroxy
6998
CHEBI:16008
2-hydroxybenzaldehyde
C1=CC=C(C(=C1)C=O)O
Boiling Point 197.0°C
Refractive Index 1.5720 to 1.5740
Flash Point 76°C
Water 0.5% Max. (K.F.)
Infrared Spectrum Authentic
Density 1.1600g/mL
Color Yellow
Packaging Glass bottle
Melting Point -7.0°C
Quantity 1kg
Specific Gravity 1.16
Assay Percent Range 99%
Linear Formula 2-(HO)C6H4CHO
Beilstein 08,31
Merck Index 15,8462
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 122.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Salicylaldehyde
Show More Show Less

Danger

Recommended Storage : product darkens during storage