Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Salicylaldehyde hydrazone, 98%

Click to view available options
Quantity:
5g
25g
100g
3291-00-7
C7H8N2O
136.154
MFCD00007626
RHPIFAMEPPCHGX-UHFFFAOYSA-N
salicylaldehyde hydrazone,benzaldehyde, 2-hydroxy-, hydrazone,2-methanehydrazonoylphenol,salicylic aldehyde hydrazone,1-salicylidenehydrazine,6-hydrazinylmethylidene cyclohexa-2,4-dien-1-one,acmc-1ap22,myunwhtzyxucik-uhfffaoysa-n,benzaldehyde,2-hydroxy-, hydrazone
6746399
6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
C1=CC(=CNN)C(=O)C=C1
3291-00-7
C7H8N2O
136.154
MFCD00007626
RHPIFAMEPPCHGX-UHFFFAOYSA-N
salicylaldehyde hydrazone,benzaldehyde, 2-hydroxy-, hydrazone,2-methanehydrazonoylphenol,salicylic aldehyde hydrazone,1-salicylidenehydrazine,6-hydrazinylmethylidene cyclohexa-2,4-dien-1-one,acmc-1ap22,myunwhtzyxucik-uhfffaoysa-n,benzaldehyde,2-hydroxy-, hydrazone
6746399
6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
C1=CC(=CNN)C(=O)C=C1
Quantity 100g
Melting Point 95°C to 98°C
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material Salicylaldehyde hydrazone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA