Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylaldoxime, 98%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
Specifications
Melting Point | 56°C to 59°C |
Sulfated Ash | 0.1% max. |
Infrared Spectrum | Authentic |
Color | Bordeaux to Purple-Red |
Water | 0.5% max. (K.F.) |
Packaging | Glass bottle |
Quantity | 100g |
Appearance | (1% soln. in chloroform) clear |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | 2-(HO)C6H4CH=NOH |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 202-353-8