Alfa Aesar™ Salicylanilide, 98%

Manufacturer: Alfa Aesar™ A1033536

Quantity 500g

 View more versions of this product

Catalog No. AAA1033536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87-17-2
    Molecular Formula C13H11NO2
    Formula Weight 213.24
    MDL Number MFCD00002212
    Merck Index 14,8330
    Synonym 2-Hydroxybenzanilide
    Chemical Name or Material Salicylanilide

    Specifications

    Formula Weight 213.24
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Synonym 2-Hydroxybenzanilide
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00002212
    Merck Index 14,8330
    Health Hazard 1 Warning
    Melting Point 136° to 138°C
    Molecular Formula C13H11NO2
    CAS 87-17-2
    Quantity 500g
    Chemical Name or Material Salicylanilide
    Assay 98%

    Description

    Applications
    It is employed as a drug used to treat rheumatoid arthritis and has principally analgesic, antipyretic and anti-inflammatory actions. Non-narcotic analgesics do not bind to opioid receptors. It is used mainly in medicated soaps.

    Solubility
    Soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit