Alfa Aesar™ Salicylhydroxamic acid, 99%

Manufacturer: Alfa Aesar™ A1152822

Quantity 100g

 View more versions of this product

Catalog No. AAA1152822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89-73-6
    Molecular Formula C7H7NO3
    Formula Weight 153.14
    MDL Number MFCD00002110
    Beilstein 1210520
    Merck Index 14,8331
    Synonym 2-Hydroxybenzohydroxamic acid
    Chemical Name or Material Salicylhydroxamic acid

    Specifications

    Formula Weight 153.14
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00002110
    CAS 89-73-6
    Synonym 2-Hydroxybenzohydroxamic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈170°C (dec.)
    Quantity 100g
    RTECS Number VO6870000
    Molecular Formula C7H7NO3
    Merck Index 14,8331
    Beilstein 1210520
    Chemical Name or Material Salicylhydroxamic acid
    Assay 99%

    Description

    Applications
    It is widely used for a variety of roles in biology and medicine as a chelating therapy. It inhibits bacterial or fungi growth by interfering with iron uptake. It is also active as a inhibitor of enzyme involved in tumour growths. These compounds are used as extractants for the separation of valuable metals or minerals including copper, lead, gold, other rare earth metals and kaolin. Hydroxamic acids are also used to produce corresponding isocyanates by dehydration followed by rearrangement reaction. It is also used as a common ligand utilized in the synthesis of metallacrowns.

    Solubility
    It is soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong bases. Stable under normal conditions.

    GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit