Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid, 99%, Thermo Scientific™

Click to view available options
Quantity:
250g
500g
2500g
69-72-7
C7H6O3
138.12
MFCD00002439
YGSDEFSMJLZEOE-UHFFFAOYSA-N
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
338
CHEBI:16914
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O
69-72-7
C7H6O3
138.12
MFCD00002439
YGSDEFSMJLZEOE-UHFFFAOYSA-N
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
338
CHEBI:16914
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.

Solubility
Soluble in carbon terachloride, benzene, propanol, absolute ethanol, acetone, toluene and ether. Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases, iodine, iron and iron salts.
Beilstein 774890
Merck Index 14,8332
Solubility Information Soluble in carbon terachloride,benzene,propanol,absolute ethanol,acetone,toluene and ether. Slightly soluble in water.
Density 1.443
Formula Weight 138.12
Percent Purity 99%
Melting Point 158°C to 160°C
Boiling Point 211°C (20mmHg)
Quantity 250g
Flash Point 157°C (315°F)
Sensitivity Light sensitive
Chemical Name or Material Salicylic acid
Show More Show Less

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c

H302-H318-H361d

EINECSNumber : 200-712-3

RTECSNumber : VO0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only