Alfa Aesar™ Salicylic acid, ACS, 99+%

Manufacturer: Alfa Aesar™ 3078222

Quantity 100g

 View more versions of this product

Catalog No. AA3078222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69-72-7
    Formula Weight 138.12
    MDL Number MFCD00002439
    Beilstein 774890
    Merck Index 14,8332
    Synonym 2-Hydroxybenzoic acid
    Chemical Name or Material Salicylic acid
    Grade ACS Grade

    Specifications

    Formula Weight 138.12
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Soluble in alcohol, acetone, ether; Slightly soluble in cold water; More soluble in boiling water; Water solubility increased by Na3PO4, alkali acetates or alkali citrates
    Boiling Point 211°C (20mmHg)
    Quantity 100g
    Synonym 2-Hydroxybenzoic acid
    Health Hazard 2 GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Melting Point 158° to 160°C
    Physical Form Powder
    MDL Number MFCD00002439
    Density 1.443g/mL
    TSCA TSCA
    CAS 69-72-7
    Beilstein 774890
    Flash Point 157° (315°F)
    Merck Index 14,8332
    Health Hazard 1 Danger
    RTECS Number VO0525000
    Chemical Name or Material Salicylic acid
    Assay ≥99%
    Grade ACS Grade

    GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit