Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

69-72-7
C7H6O3
138.122
YGSDEFSMJLZEOE-UHFFFAOYSA-N
338
CHEBI:16914
2-hydroxybenzoic acid
C1=CC=C(C(=C1)C(=O)O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 138.12
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.44kg
Quantity 1kg
Flash Point 157.00°C
Chemical Name or Material Salicylic Acid
Grade Baker Analyzed™ ACS Reagent