Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Crystal, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle

S1005, 69-72-7, C7H6O3

Spectrum™ Salicylic Acid, Crystal, Reagent, ACS is an alpha-hydroxybenzoic acid and a colorless crystalline acid that is organic and commonly used in organic synthesis. It can be found occurring naturally in unripe fruits and vegetables and most commonly in the white willow tree. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

CAS 69-72-7
CAS Max % 1
Molecular Formula C7H6O3
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1O
IUPAC Name 2-hydroxybenzoic acid
Molecular Weight (g/mol) 138.12
Percent Purity 99%
Grade ACS
Heavy Metals (as Pb) 5 ppm
Iron (Fe) 2 ppm
Packaging Amber Glass Bottle
Residue after Ignition 0.0001
Sulfate 3e-005
Melting Point 158°C to 161°C
Quantity 500 g
Show More Show Less