Salicylic Acid (Crystalline/USP), Fisher Chemical

Packaging Fiber Drum
Quantity 12kg

 View more versions of this product

Catalog No. A275-12

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 69-72-7
    Physical Form Solid
    Molecular Formula C7H6O3
    Linear Formula 2-HOC6H4COOH
    Formula Weight 138.12
    MDL Number MFCD00002439
    Synonym 2-Hydroxybenzoic Acid
    Chemical Name or Material Salicylic Acid
    Assay 99.5 to 101.0% (USP)
    Grade USP

    Specifications

    Melting Point 158°C
    Boiling Point 211°C
    Formula Weight 138.12
    Quantity 12kg
    pH 2.4
    Color Off-white
    Physical Form Solid
    Vapor Pressure 0.3mbar at 95°C
    Heavy Metals (as Pb) 20ppm max. (USP)
    Identification Pass Test (USP)
    Melting Temp_End 161°C max. (USP)
    Sulfate 0.02% max. (USP)
    Total Impurity 0.2% max. (USP)
    Chloride 0.014% max. (USP)
    MDL Number MFCD00002439
    ChemAlert Storage Symbol Gray
    Synonym 2-Hydroxybenzoic Acid
    Health Hazard 1 SIGNAL WORD: DANGER!
    Ignition Residue 0.05% max. (USP)
    Loss on Drying 0.5% max. (USP)
    Melting Temp_Begin 158°C min. (USP)
    Packaging Fiber Drum
    Salicylic Acid Related Compound A 0.1% max. (USP)
    Molecular Formula C7H6O3
    Linear Formula 2-HOC6H4COOH
    Health Hazard 3 Emergency Overview
    Harmful if swallowed. Irritating to eyes, respiratory system and skin.
    NFPA
    Health: 2
    Flammability: 1
    Instability: 0
    Other Impurities 0.05% max. (USP)
    Phenol Relative Retention Time 0.02% max. (USP)
    Residual Solvents Meets Requirements (USP)
    Salicylic Acid Related Compound B 0.05% max.
    CAS 69-72-7
    Chemical Name or Material Salicylic Acid
    Assay 99.5 to 101.0% (USP)
    Grade USP
     

    Emergency Overview
    Harmful if swallowed. Irritating to eyes, respiratory system and skin.
    NFPA
    Health: 2
    Flammability: 1
    Instability: 0

    SIGNAL WORD: DANGER!

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit