Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid (Crystalline/USP), Fisher Chemical

Packaging Poly Pail
Quantity 2.5kg

 View more versions of this product

Catalog No. A275-212

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

69-72-7
C7H6O3
138.12
2-Hydroxybenzoic Acid
99.5 to 101.0% (USP)
Solid
2-HOC6H4COOH
MFCD00002439
Salicylic Acid
USP

Specifications

69-72-7
0.014% max. (USP)
2-HOC6H4COOH
MFCD00002439
20ppm max. (USP)
0.05% max. (USP)
158°C min. (USP)
0.05% max. (USP)
0.02% max. (USP)
0.1% max. (USP)
0.02% max. (USP)
0.3mbar at 95°C
2.4
211°C
Salicylic Acid
USP
Solid
C7H6O3
138.12
2-Hydroxybenzoic Acid
Pass Test (USP)
0.5% max. (USP)
161°C max. (USP)
Poly Pail
Meets Requirements (USP)
0.05% max.
0.2% max. (USP)
158°C
Off-white
2.5kg
99.5 to 101.0% (USP)

Safety and Handling

SDS & Safety Documents

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit