Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Powder, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
69-72-7
C7H6O3
138.12
YGSDEFSMJLZEOE-UHFFFAOYSA-N
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O
69-72-7
C7H6O3
138.12
YGSDEFSMJLZEOE-UHFFFAOYSA-N
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O

SA130, 69-72-7, C7H6O3

Spectrum™ Salicylic Acid, Powder, USP is a topical application used to treat skin conditions that involve scaling or overgrowth of skin cells such as psoriasis, dandruff, corns, calluses, and warts. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 98 to 102%
Grade USP
Ignition Residue 0.0005
Loss on Drying 0.005
Packaging Poly Pail
Salicylic Acid Related Compound A 0.001
Salicylic Acid Related Compound B 0.0005
Sulfate 0.0002
Total Impurity 0.002
Quantity 2.5 kg
Show More Show Less