Alfa Aesar™ Salicylsalicylic acid, 98%

Manufacturer: Alfa Aesar™ B2244414

Quantity 25g

 View more versions of this product

Catalog No. AAB2244414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 552-94-3
    Molecular Formula C14H10O5
    Formula Weight 258.23
    MDL Number MFCD00020252
    Synonym 2-Hydroxybenzoic acid 2-carboxyphenyl ester
    Chemical Name or Material Salicylsalicylic acid

    Specifications

    Formula Weight 258.23
    TSCA TSCA
    Molecular Formula C14H10O5
    MDL Number MFCD00020252
    Synonym 2-Hydroxybenzoic acid 2-carboxyphenyl ester
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 132° to 138°C
    Quantity 25g
    RTECS Number DH2080000
    CAS 552-94-3
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Salicylsalicylic acid
    Assay 98%

    Description

    Applications
    Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

    Solubility
    Hydrolyzes slowly in water.

    Notes
    Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit