Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Salicylsalicylic acid, 98%

Click to view available options
Quantity:
25g
100g
500g
552-94-3
C14H10O5
258.229
MFCD00020252
WVYADZUPLLSGPU-UHFFFAOYSA-N
salsalate,sasapyrine,salicylsalicylic acid,disalicylic acid,disalcid,sasapyrinum,saloxium,2-carboxyphenyl salicylate,salina,diacesal
5161
CHEBI:9014
2-(2-hydroxybenzoyl)oxybenzoic acid
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O
552-94-3
C14H10O5
258.229
MFCD00020252
WVYADZUPLLSGPU-UHFFFAOYSA-N
salsalate,sasapyrine,salicylsalicylic acid,disalicylic acid,disalcid,sasapyrinum,saloxium,2-carboxyphenyl salicylate,salina,diacesal
5161
CHEBI:9014
2-(2-hydroxybenzoyl)oxybenzoic acid
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O
Applications
Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 500g
Melting Point 132°C to 138°C
Formula Weight 258.23
Percent Purity 98%
Chemical Name or Material Salicylsalicylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DH2080000

TSCA : TSCA