Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Saline 0.85% Solution Tubes
Reduce diluent preparation time using convenient, pre-filled and ready-to-use 0.85% saline tubes.
Supplier: Thermo Scientific™ R07142
Description
Save time by removing the need to manually prepare isotonic diluents using Thermo Scientific™ Saline 0.85% Solution Tubes.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.- Easily adjust inoculum density with a clear tube.
- Enhance sample security while promoting safe handling with a reliable screw cap on each tube
- Achieve versatility and flexibility with multiple fill volumes available
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Clear liquid colorless | |
Saline 0.85% Solution Tubes | |
United States | |
9.9 mL | |
20/Pk. |
2 to 25°C | |
16mm x 125mm Tube | |
Buffer Solution | |
Glass tube with screw cap |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction