Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents
$108.00 - $181.00

Specifications
Chemical Name or Material | Saline-Sodium Phosphate-EDTA |
---|---|
Name Note | 20X Solution |
CAS | 7647-14-5, 7558-80-7, 60-00-4, 7732-18-5 |
Synonym | SSPE Buffer |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP1328-1
|
1L | Nalgene Poly Bottle |
Each for $108.00
Case of 6 EA for $516.12
Order in Bulk
Request bulk or custom formats |
|
BP1328-4
|
4L | Poly CUBE |
Each for $181.00
Order in Bulk
Request bulk or custom formats |
|
Description
SalineSodium PhosphateEDTA (SSPE) is used in bacterial screening and hybridization procedures.Specifications
Saline-Sodium Phosphate-EDTA | |
20X Solution | |
7647-14-5, 7558-80-7, 60-00-4, 7732-18-5 | |
SSPE Buffer |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title