Learn More
LaMotte Salinity Test
Test sea water, marine estuaries, aquariums and brine solutions.
$84.25
Specifications
Range | 0 to 40ppt |
---|---|
For Use With (Equipment) | Direct reading titrator |
Includes | 3 reagents |
No. of Tests | 50.0 |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S450911
![]() ![]() |
LaMotte
745902 |
Salinity Water Test Kit |
Each for $84.25
|
|
Description
- Uses a direct-reading titrator
- Conduct accurate salt water analysis
- Measures 0 to 20ppt salinity in 0.4ppt increments
- Range can be extended to 0 to 40ppt
- Utilizes modified APHA argentometric method
- Contains three reagents
- Enough materials for 50 tests at 20ppt
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
0 to 40ppt | |
Direct reading titrator | |
3 reagents | |
50.0 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.