Learn More
Tocris Bioscience™ Salirasib Available on GSA/VA Contract for Federal Government customers only.
Ras inhibitor; also induces autophagy
Manufacturer:  Tocris Bioscience™ 4989/10
 View more versions of this product
Catalog No. 49-891-0R
Add to cart
This item is not returnable. View return policy
Description
Description
Inhibitor of active Ras proteins. Displaces active Ras from the plasma membrane; impairs downstream signaling and inhibits proliferation of endometrial carcinoma cells. Also facilitates Ras degradation. Shown to induce autophagy in several human cancer cell lines.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Salirasib
>98
10mg
Ras GTPase Inhibitors
358.54