Learn More
Honeywell Salisbury™ Disposable Face Shield
Utilizes nanotechnology to provide a clearer, more transparent window for improved visibility
Supplier: Honeywell Salisbury™ AS1200HATPPC
This item is not returnable.
View return policy
Description
face shield unit includes ventilation ribs that allow a natural convection airflow through the top of the face shield; reduces fogging of the visor, lowers CO2 levels and improves comfort for the user.
- ATPV rating: 12cal/cm2
- Weight balancing arc flash protection face shield
- Stows in a balanced, compact position, centered over the top of the hardhat when not needed
- The shield/window is designed to be easily replaced without tools
Optional Accessories (contact your Fisher Safety Representative):
- Cold weather anti-fog inner lens and attachable task lights; includes an air spaced design that decreases fogging in cold weather
- Attachable task lights; held in place by adjustable flashlight clip on both sides of face shield
Specifications
Premium Polycarbonate Lens, Arc Flash Protection Face Shield, Hard Hat | |
Face Shield | |
Meets ANSI Z87.1 standards; Tested to ASTM F2187 specifications |