Learn More
Honeywell Salisbury™ Arc Flash Protection Hoods
Supplier: Honeywell Salisbury™ AFHOOD10
This item is not returnable.
View return policy
Description
Specifications
Universal | |
White | |
10 cal/cm2 |
Nomex, Lenzing | |
Must be worn in combination with face shield, AS1000 series faceshield/hard hat/chin cup unit | |
1 Ea. |