Salisbury Arc Flash Protection Hoods

Manufacturer: WH Salisbury & Co. AFHOOD20

 View more versions of this product

Catalog No. 19-139-702

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Hood Material P84, Lenzing™, Kevlar™
    Quantity Each
    Material P84, Lenzing™, Kevlar™
    Size Universal
    Arc Flash Rating 20 Calories/cm2
    For Use With (Equipment) Must be worn in combination with face shield, AS1000 series faceshield/hard hat/chin cup unit
    Color Beige

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit