Learn More
Salisbury™ Class 3 High Voltage Linemen's Gloves
Contrast between the thin outer color against the inner color makes inspecting for cuts and tears easier when the glove is inflated or stretched
Manufacturer:  WH Salisbury & Co. E318RB/10
 View more versions of this product
Catalog No. 19-816-082
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Straight
Rubber, natural
26500V
Black,Red
18 in.
3
utility lineman, telecommunications, HVAC, field service and automotive technicians
10
ASTM D 120, IEC EN 60903
Red
45.7cm