Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Class O Low-Voltage Lineman's Gloves

Ideal for use in meter installation and maintenance

$60.64 - $157.20

Specifications

Arc Flash Yes
Certifications/Compliance ASTM D 120, IEC EN 60903
Classification 0
Material Rubber
For Use With (Application) electrical contracting and power generation
View More Specs

Products 12
Catalog Number Mfr. No. Color Length (English) Length (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Length (English) Length (Metric) Size Price Quantity & Availability  
11-394-22A View Documents Honeywell Salisbury™
E011BY/8
Yellow 11 in. 27.9 cm 8 Pair for $157.20
Add to cart
 
11-394-22B View Documents Honeywell Salisbury™
E011BY/9
Yellow 11 in. 27.9 cm 9 Pair for $157.20
Add to cart
 
11-395-40A View Documents Honeywell Salisbury™
E011B/8
Black 11 in. 27.9 cm 8 Pair for $60.64
Add to cart
 
11-395-40B View Documents Honeywell Salisbury™
E011B/9
Black 11 in. 27.9 cm 9 Pair for $139.70
Add to cart
 
11-395-40C View Documents Honeywell Salisbury™
E011B/9H
Black 11 in. 27.9 cm 9.5 Pair for $135.70
Add to cart
 
11-395-40D View Documents Honeywell Salisbury™
E011B/10
Black 11 in. 27.9 cm 10 Pair for $153.20
Add to cart
 
11-395-40F View Documents Honeywell Salisbury™
E011B/11
Black 11 in. 27.9 cm 11 Pair for $153.20
Add to cart
 
11-395-40G View Documents Honeywell Salisbury™
E011BL/11
Blue 11 in. 27.9 cm 11.5 Pair for $135.70
Add to cart
 
19-816-054 View Documents Honeywell Salisbury™
E011R/9H
Red 11 in. 27.9 cm 9.5 Pair for $140.80
Add to cart
 
19-025-612 View Documents Honeywell Salisbury™
E011R/10H
Red 11 in. 27.9 cm 10.5 Pair for $143.00
Add to cart
 
11-394-22C View Documents Honeywell Salisbury™
E011BY/10
Yellow 11 in. 27.9 cm 10 Pair for $157.20
Add to cart
 
19-821-347 View Documents Honeywell Salisbury™
E014R/8
Red 14 in. 35.5 cm 8 Pair for $144.10
Add to cart
 
Description

Description

Ideal for use in meter installation and maintenance

  • Made from natural rubber with a special formulation that provides dexterity and protection beyond the wrist
  • Available in 11 and 14 in. (27.9 and 35.6cm) lengths
  • Tested at 5kVAC and listed for a maximum voltage of 1000VAC in accordance with ASTM™ D 120-87
  • Sold by the pair

Specifications

Specifications

Yes
0
electrical contracting and power generation
1000VAC, 1500VDC
ASTM D 120, IEC EN 60903
Rubber
Flared
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit