Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ G99 Portable Glove Inflator Kit - Replacement Parts

The G99 Inflator Kit provides a quality means of inspecting gloves in the field. Replacement parts ensure accurate operation and help to extend product lifetime.

$7.10 - $18.75

Specifications

For Use With (Equipment) G99 Portable Glove Inflator Kit
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19-138-421
View Documents Promotion Details
Honeywell Salisbury™
G99V
Check Valve
Each for $7.10
Add to cart
 
19-138-420
View Documents Promotion Details
Honeywell Salisbury™
G99S
Nylon Strap
Each for $10.76
Add to cart
 
19-138-418
View Documents Promotion Details
Honeywell Salisbury™
G99B
Storage Bag
Each for $18.75
Add to cart
 
Description

Description

  • The G99 is a simple, easy to use, portable glove inflator
  • Provides a quality means of inspecting gloves in the field
  • Glove is secured to the inflator by a nylon strap, and fastened with a hook and pile closure
  • Inflation is accomplished by pumping the bellows of the inflator against any surface
  • Type I gloves should be inflated no more than 1.5x their normal size
  • Type II Salcor™ gloves should be inflated no more than 1.25x their normal size
  • Replacement parts help ensure that the pump is working properly whenever and wherever it is needed
Specifications

Specifications

G99 Portable Glove Inflator Kit
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit