Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Insulated Class 0 Glove Kits

Proper storage to extend the service life of linemen's gloves and sleeves

$259.50 - $275.00

Specifications

Arc Flash Yes
Classification 0
Length (English) 14 in.
Length (Metric) 35.5 cm
For Use With (Application) storage of insulating Rubber gloves and Leather protectors
View More Specs

Products 22
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
19-139-770 View Documents Honeywell Salisbury™
GK014B/7
Black 7 Each for $259.50
Add to cart
 
19-139-771 View Documents Honeywell Salisbury™
GK014B/8
Black 8 Each for $259.50
Add to cart
 
19-139-775 View Documents Honeywell Salisbury™
GK014B/12
Black 12 Each for $259.50
Add to cart
 
19-139-760 View Documents Honeywell Salisbury™
GK011BL/9
Blue 9 Each for $270.00
Add to cart
 
19-139-759 View Documents Honeywell Salisbury™
GK014BL/8
Blue 8 Each for $268.50
Add to cart
 
19-139-761 View Documents Honeywell Salisbury™
GK014BL/10
Blue 10 Each for $268.50
Add to cart
 
19-139-762 View Documents Honeywell Salisbury™
GK014BL/11
Blue 11 Each for $268.50
Add to cart
 
19-139-763 View Documents Honeywell Salisbury™
GK014BL/12
Blue 12 Each for $268.50
Add to cart
 
19-139-752 View Documents Honeywell Salisbury™
GK014R/7
Red 7 Each for $267.50
Add to cart
 
19-139-755 View Documents Honeywell Salisbury™
GK014R/10
Red 10 Each for $267.50
Add to cart
 
19-139-753 View Documents Honeywell Salisbury™
GK014R/8
Red 8 Each for $267.50
Add to cart
 
19-139-754 View Documents Honeywell Salisbury™
GK014R/9
Red 9 Each for $272.00
Add to cart
 
19-139-756 View Documents Honeywell Salisbury™
GK014R/11
Red 11 Each for $267.50
Add to cart
 
19-139-757 View Documents Honeywell Salisbury™
GK014R/12
Red 12 Each for $267.50
Add to cart
 
19-139-765 View Documents Honeywell Salisbury™
GK014Y/8
Yellow 8 Each for $275.00
Add to cart
 
19-139-766 View Documents Honeywell Salisbury™
GK014Y/9
Yellow 9 Each for $275.00
Add to cart
 
19-139-767 View Documents Honeywell Salisbury™
GK014Y/10
Yellow 10 Each for $265.50
Add to cart
 
19-139-768 View Documents Honeywell Salisbury™
GK014Y/11
Yellow 11 Each for $275.00
Add to cart
 
19-139-769 View Documents Honeywell Salisbury™
GK014Y/12
Yellow 12 Each for $275.00
Add to cart
 
19-139-774 View Documents Honeywell Salisbury™
GK014B/11
Black 11 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-139-773 View Documents Honeywell Salisbury™
GK014B/10
Black 10 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-139-772 View Documents Honeywell Salisbury™
GK014B/9
Black 9 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Kits are 35.5cm (14 in.) in length and made of rubber
  • Available in a variety of colors

Specifications

Specifications

Yes
14 in.
storage of insulating Rubber gloves and Leather protectors
0
35.5 cm
Pair of Rubber gloves, Leather protectors and sized glove bag
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit