Salisbury™ Insulated Class 0 Glove Kits

Proper storage to extend the service life of linemen's gloves and sleeves

Manufacturer: WH Salisbury & Co. GK014B/10

 View more versions of this product

Catalog No. 19-139-773

  Check Availability
Add to Cart

    Specifications

    Includes Pair of Rubber gloves, Leather protectors and sized glove bag
    For Use With (Application) storage of insulating Rubber gloves and Leather protectors
    Length (English) 14 in.
    Length (Metric) 35.5cm
    Size 10
    Color Black
    Classification 0
    Arc Flash Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit