Learn More
Salisbury Insulated Rescue Hook
Used to withdraw injured workers from hazardous locations
Manufacturer:  WH Salisbury & Co. 24401
 View more versions of this product
Catalog No. 19-138-495
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rescue hook
Coated, heat treated body hook with 18 in. opening
180cm
4.5 lb.
Confined spaces, in vaults or near electrical cabinets and switch gear
72 in.
Foam Filled Fiberglass
2kg