Salisbury LP Series Glove Protectors

Offer excellent comfort and protection

Manufacturer: WH Salisbury & Co. LP10/10H

 View more versions of this product

Catalog No. 19-025762

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Rubber insulating gloves
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Size 10.5
    Color Orange
    Material Vinyl, Cow Skin

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit