Learn More
Honeywell Salisbury™ LP Series Glove Protectors
Offer excellent comfort and protection
Manufacturer:  Honeywell Salisbury™ LP10/9
 View more versions of this product
Catalog No. 11-393-92B
Add to cart
Description
Description
Manufactured from top grain cowhide or goatskin.
  • Designed to fit with Salisbury Class 00 and Class 0 insulating rubber gloves to maximize wear-of-life
  • Palm side of cuff is constructed of tough leather for superior wear resistance
  • 10 in. long (25cm) protector has a shirred elastic back to secure to electrical gloves
  • 12 in. (30cm), 14 in. (36cm), and 16 in. (40.6cm) long protectors have orange vinyl-coated fabric back cuff for high visibility and adjustable straps fitted with nonmetallic buckles
  • All styles feature tailored leather strip reinforcement to protect thumb crotch
Compliance
Manufactured to meet ASTM™ F696
Specifications
Specifications
Safety
Cowhide Leather, Vinyl
Orange
25.4 cm
Rubber insulating gloves
9
10 in.