Learn More
Honeywell Salisbury PRO-HOOD Arc Flash Protective Hoods
Lighter tint lenses for better visual light transmission
$405.00
Specifications
Certifications/Compliance | ASTM, F2178, NFPA 70E™, ASTM F1506 |
---|---|
Length (English) | 18 in. |
Length (Metric) | 45.72 cm |
Hood Material | Cotton, Nylon |
Includes | Universal hard hat bracket |
Catalog Number | Mfr. No. | Arc Flash Rating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Arc Flash Rating | Price | Quantity & Availability | |||||
19-138-110
|
Honeywell Salisbury
FH11BL |
12 cal/cm2 |
Each for $405.00
|
|
Description
- 11 cal/cm2 to 100 cal/cm2 ATPV ratings
- Made of arc-flash-resistant Indura Ultra Soft material and sewn with Nomex thread
- 100 cal/cm2 is made from material layers of TuffWeld and Q/9
- Arc-rated 10 x 20 in. antifogging replaceable lens
- Slotted bracket
- Hook-and-pile tabs to attach PRO-AIR belt-mounted compact air system
- One size fits all
Compliance
Meet current ASTM F2178 and NFPA 70E standards and the flammability requirements for ASTM F1506.
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40cal/cm2 should be avoided becaouse of blast hazards caused by arc flash.
Specifications
ASTM, F2178, NFPA 70E™, ASTM F1506 | |
45.72 cm | |
Universal hard hat bracket | |
1 Ea. |
18 in. | |
Cotton, Nylon | |
Arc-rated Cotton, Nylon | |
Universal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.