Salisbury™ PRO-HOOD™ Arc Flash Protective Hoods
Lighter tint lenses for better visual light transmission
Manufacturer: WH Salisbury & Co. FH40GY
 View more versions of this product
Catalog No. 19-138-113
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM, F2178, NFPA 70E™, ASTM F1506
    Hood Material Cotton, Nylon
    Quantity Each
    Includes Universal hard hat bracket
    Material Arc rated Cotton, Nylon
    Size Universal
    Arc Flash Rating 40 Calories/cm2
    Length (English) 18 in.
    Length (Metric) 45.72cm
    Color Gray
  • Description & Specifications
Related Products
Product Content Feedback