Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ PRO-WEAR™ 10 cal/cm2 Faceshield with Hardhat

Arc flash faceshield with hardhat and chin cup

$144.10

Specifications

For Use With (Equipment) Salisbury PRO-SHIELD Face Shield System
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
19-138-104
View Documents
Honeywell Salisbury™
AS BAG
Storage Bag Drawstring closure and Fleece interior
Each of 1 for $144.10
Only null left
Add to Cart
 
Description

Description

  • 10 cal/cm2 ATPV rating when used with included chin cup (8 cal/cm2 without)
  • Extra-light tint faceshield is 0.06 in. thick and has large 7 1/2 × 20 in. viewing area
  • Faceshield features 70% visual light transmission, absorbs > 99.9% of harmful UV radiation, and resists fogging
  • Orange front-brim hardhat features four-point ratcheting nylon suspension is ANSI 289.1-2003, Type I, Class E dielectrically rated
  • Optional canvas storage bag available with fleece interior and drawstring closure to protect faceshield and hardhat

Compliance

Tested to ASTM F2178 specifications

Specifications

Specifications

Salisbury PRO-SHIELD Face Shield System
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.