Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Salisbury PRO-WEAR Arc Flash Personal Protection Equipment Kits
ATPV Rating: 40 cal/cm2
Specifications
Arc Flash Rating | 40 Calories/cm2 |
---|---|
Certifications/Compliance | ASTM F1506, NFPA 70E™, CE 0120 |
Description | Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-138-167E
|
Honeywell Salisbury
SK402XL |
2X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-138-167C
|
Honeywell Salisbury
SK40L |
Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-138-167B
|
Honeywell Salisbury
SK40M |
Medium |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-138-167F
|
Honeywell Salisbury
SK403XL |
3X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-138-167D
|
Honeywell Salisbury
SK40XL |
X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Kit contains an arc flash coat, bib overalls, PRO-HOOD, hardhat, safety glasses and SKBAG; does not include gloves. PRO-WEAR 40 Cal/cm2 products are made from Milliken Amplitude material to be flame resistant for the life of the garment.
Arc Flash Coat
- Constructed of extremely lightweight material
- Coat has expansion back design for added comfort
- Flame-resistant cotton; sewn with Nomex thread
- Dual-stage front closure with high-temperature plastic zipper
- Hook-and-pile storm flap for added protection
- Flame-resistant Nomex wristlets
- Color: Gray
- Bib overalls have heavy-duty suspenders for a secure fit
- Adjustable hook and pile waist straps
- Hood has arc rated, 10 x 20 in. antifogging replaceable lens
- For use with insulating rubber gloves and leather protectors (not included)
Improved lens clarity
Certifications
Hazard risk Category 4.
Compliance
Meet NFPA 70E and ASTM F1506 standards.
Specifications
40 Calories/cm2 | |
ASTM F1506, NFPA 70E™, CE 0120 | |
Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title